2-Aminopurine, 98%
2-Aminopurine, 98%
2-Aminopurine, 98%
Thermo Scientific Chemicals

2-Aminopurine, 98%

A protein kinase R inhibitor | CAS: 452-06-2 | C5H5N5 | 135.13 g/mol
数量:
250 mg
500 mg
1 g
製品番号(カタログ番号) J64919.MD
または、製品番号J64919-MD
価格(JPY)
-
見積もりを依頼する
数量:
250 mg
一括またはカスタム形式をリクエストする
仕様
CAS452-06-2
ChEBICHEBI:479072
化学物質名または材質2-Aminopurine
色White to Yellow
EINECS番号207-197-4
さらに表示
A protein kinase R inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

よくあるご質問(FAQ)

What is the solubility of 2-Aminopurine 98%?

We have the following information regarding solubility for 2-Aminopurin 98%:
- Soluble in a formic acid and water solution (1:1 v/v) at 50 mg/ml by heating.
- Soluble in a glacial acetic acid and PBS solution (200:1) by heating at 60 degrees C for an 150 mM solution. This solution can then be aliquoted and stored frozen. The aliquots should be thawed, heated and mixed before usage.

What is the molecular weight of 2-Aminopurine 98%?

The molecular weight is 135.13 g/mol (this product is the non-salt form, CAS 452-06-2).