Ghrelin, Human
Thermo Scientific Chemicals

Ghrelin, Human

 
製品番号(カタログ番号)数量
J64943.MCR
または、製品番号J64943-M^
1 mg
製品番号(カタログ番号) J64943.MCR
または、製品番号J64943-M^
価格(JPY)
-
数量:
1 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS258279-04-8
Appearance (Color)White
FormPowder
Assay (HPLC)≥95.0%
CommentCounter Ion: Reported
The GHS-R (growth hormone secretagogue receptor) potently stimulates growth hormone release from the pituitary gland in vivo and in vitro. It stimulates food intake and transduces signals to hypothalamic regulatory nuclei that control energy homeostasis. Ghrelin (human) is an activator of GHS-R1a.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
組成および保存条件
-30°C to -10°C