5-Methyl-2'-deoxycytidine, 99%
5-Methyl-2'-deoxycytidine, 99%
5-Methyl-2'-deoxycytidine, 99%
Thermo Scientific Chemicals

5-Methyl-2'-deoxycytidine, 99%

CAS: 838-07-3 | C10H15N3O4 | 241.247 g/mol
製品番号(カタログ番号)数量
J65209.03
または、製品番号J65209-03
1 g
製品番号(カタログ番号) J65209.03
または、製品番号J65209-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS838-07-3
ChEBICHEBI:47876
化学物質名または材質5-Methyl-2'-deoxycytidine
White
EINECS番号212-655-1
さらに表示
5-Methyl-2'-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-2′-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditio
RUO – Research Use Only

General References:

  1. Brian C. Froehler.; Daniel J. Ricca. Triple-helix formation by oligodeoxynucleotides containing the carbocyclic analogs of thymidine and 5-methyl-2'-deoxycytidine. J. Am. Chem. Soc. 1992, 114, (21), 8320-8322.
  2. Kuei-Ying Lin.; Robert J. Jones.; Mark Matteucci. Tricyclic 2'-Deoxycytidine Analogs: Syntheses and Incorporation into Oligodeoxynucleotides Which Have Enhanced Binding to Complementary RNA. J. Am. Chem. Soc. 1995, 117, (13), 3873-3874.