8-Anilinonaphthalene-1-sulfonic acid, 95%
8-Anilinonaphthalene-1-sulfonic acid, 95%
8-Anilinonaphthalene-1-sulfonic acid, 95%
8-Anilinonaphthalene-1-sulfonic acid, 95%
Thermo Scientific Chemicals

8-Anilinonaphthalene-1-sulfonic acid, 95%

Fluorescent probe for proteins on polyacrylamide gels | CAS: 82-76-8 | C16H13NO3S | 299.344 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) J65332.06
または、製品番号J65332-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS82-76-8
IUPAC Name8-(phenylamino)naphthalene-1-sulfonic acid
Molecular FormulaC16H13NO3S
InChI KeyFWEOQOXTVHGIFQ-UHFFFAOYSA-N
SMILESOS(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1
さらに表示
Appearance (Color)Pale grey to black or brown
FormCrystals or powder or crystalline powder
Assay by NMR≥94.0% (Proton-NMR)
Fluorescent probe for proteins on polyacrylamide gels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only