8-Anilinonaphthalene-1-sulfonic acid ammonium salt, 98%
8-Anilinonaphthalene-1-sulfonic acid ammonium salt, 98%
8-Anilinonaphthalene-1-sulfonic acid ammonium salt, 98%
8-Anilinonaphthalene-1-sulfonic acid ammonium salt, 98%
Thermo Scientific Chemicals

8-Anilinonaphthalene-1-sulfonic acid ammonium salt, 98%

Fluorescent probe for proteins on polyacrylamide gels | CAS: 28836-03-5 | C16H16N2O3S | 316.38 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) J65538.14
または、製品番号J65538-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS28836-03-5
IUPAC Nameammonium 8-(phenylamino)naphthalene-1-sulfonate
Molecular FormulaC16H16N2O3S
InChI KeyIPBNQYLKHUNLQE-UHFFFAOYSA-N
SMILES[NH4+].[O-]S(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1
さらに表示
FormPowder or crystalline powder
Assay from Supplier's CofA≥98.0% (HPLC)
Loss on Drying≤5.0%
Proton NMRConforms to structure
Appearance (Color)Straw yellow or green to dark green
ANS forms an inclusion complex with cyclodextrin. Such model systems are useful to mimic biological recognition and can be studied by measuring the change in fluorescence of free-ANS to complexed-ANS. When ANS enters the hydrophobic core of cyclodestrin, it’s fluorescence increases. It is commonly used to study molecular assemblies of surfactants and amphiphilic polymers because a blue shift of the emission maximum indicates the fluorophore is located in less polar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ANS forms an inclusion complex with cyclodextrin. Such model systems are useful to mimic biological recognition and can be studied by measuring the change in fluorescence of free-ANS to complexed-ANS. When ANS enters the hydrophobic core of cyclodestrin, it’s fluorescence increases. It is commonly used to study molecular assemblies of surfactants and amphiphilic polymers because a blue shift of the emission maximum indicates the fluorophore is located in less polar media.

Solubility
Soluble in water, 1N NaOH, and methanol.

Notes
Light & Air Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Neha Jain.; Mily Bhattacharya.; Samrat Mukhopadhyay. Kinetics of Surfactant-induced Aggregation of Lysozyme Studied by Fluorescence Spectroscopy. Journal of Fluorescence. 2011, 21, (2), 615-625.