2'-Fluoro-2'-deoxyuridine, 97%
2'-Fluoro-2'-deoxyuridine, 97%
2'-Fluoro-2'-deoxyuridine, 97%
2'-Fluoro-2'-deoxyuridine, 97%
Thermo Scientific Chemicals

2'-Fluoro-2'-deoxyuridine, 97%

CAS: 784-71-4 | C9H11FN2O5 | 246.19 g/mol
製品番号(カタログ番号) J65948.06
または、製品番号J65948-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS784-71-4
IUPAC Name1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H11FN2O5
InChI KeyUIYWFOZZIZEEKJ-XVFCMESISA-N
SMILESOC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(=O)NC1=O
さらに表示
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥96.0%
Appearance (Color)White to off-white
2'-Fluoro-2'-deoxyuridine is used as a component incorporated into therapeutic oligonucleotides to combat prostate and breast cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoro-2′-deoxyuridine is used as a component incorporated into therapeutic oligonucleotides to combat prostate and breast cancer.

Solubility
Soluble in methanol.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Charles L. Perrin. Mechanism of hydrogen exchange in amides. J. Am. Chem. Soc. 1974, 96, (17), 5628-5631.
  2. Osamu Shimomura; Ikuyoshi Tomita; Takeshi Endo. Cationic polymerization of glycidyl phenyl ether initiated by various arylsulfoniu∼ salts as new thermal latent cationic initiators. Macromol. Rapid Cammun. 1998, 19 (9), 493-497 .