D-Ribonic acid-1,4-lactone
D-Ribonic acid-1,4-lactone
D-Ribonic acid-1,4-lactone
D-Ribonic acid-1,4-lactone
Thermo Scientific Chemicals

D-Ribonic acid-1,4-lactone

CAS: 5336-08-3 | C5H8O5 | 148.11 g/mol
製品番号(カタログ番号) J66266.06
または、製品番号J66266-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5336-08-3
IUPAC Name3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one
Molecular FormulaC5H8O5
InChI KeyCUOKHACJLGPRHD-UHFFFAOYNA-N
SMILESOCC1OC(=O)C(O)C1O
さらに表示
Optical RotationSpecific optical rotation [α]: +17 to + 19º (20ºC, C=5, H2O)
FormWhite to off-white crystalline powder
Assay (GC)≥ 97%
Assay by NMRConforms structure
Residue on ignition≤ 0.2%
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only