Mezlocillin sodium salt, Thermo Scientific Chemicals
Mezlocillin sodium salt, Thermo Scientific Chemicals
Mezlocillin sodium salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mezlocillin sodium salt, Thermo Scientific Chemicals

A broad spectrum antibiotic
製品番号(カタログ番号)数量
J66306.MC
または、製品番号J66306-MC
100 mg
製品番号(カタログ番号) J66306.MC
または、製品番号J66306-MC
価格(JPY)
-
数量:
100 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS59798-30-0
IUPAC Namesodium 6-{2-[(3-methanesulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetamido}-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular FormulaC21H24N5NaO8S2
InChI KeyGTGQRSIMEUWHPA-UHFFFAOYNA-M
SMILES[Na+].CC1(C)SC2C(NC(=O)C(NC(=O)N3CCN(C3=O)S(C)(=O)=O)C3=CC=CC=C3)C(=O)N2C1C([O-])=O
さらに表示
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
Assay (unspecified)≥90.0%
pH4.5-7.5
Water Content≤6.0%
A broad spectrum antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only