Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals

D-Lactose

Catalog number: J66376.A1
1 kg, Each
Thermo Scientific Chemicals

D-Lactose

Catalog number: J66376.A1
1 kg, Each
Quantity
Catalog number: J66376.A1
also known as J66376-A1
Price (USD)

Chemical Identifiers

CAS
63-42-3
IUPAC Name
2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular Formula
C12H22O11
InChI Key
GUBGYTABKSRVRQ-UHFFFAOYNA-N
SMILES
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
Form
Powder
Assay from Suppliers CofA
≥98.0%
Appearance (Color)
White to off-white

Description

D-Lactose is used for the culture of lactic acid metabolizing bacteria, lactic acid bacteria. D-Lactose is used to identify and characterized galectins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• D-Lactose is the major sugar found in milk
• It is formed through the condensation of galactose and glucose
Application
• D-Lactose is used to identify and characterize galectins
• It is also used to culture lactic acid-metabolizing bacteria
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text