Oxacillin sodium salt monohydrate, 815^mg/mg
Oxacillin sodium salt monohydrate, 815^mg/mg
Oxacillin sodium salt monohydrate, 815^mg/mg
Thermo Scientific Chemicals

Oxacillin sodium salt monohydrate, 815^mg/mg

CAS: 7240-38-2 | C19H23N3NaO6S | 444.458 g/mol
製品番号(カタログ番号)数量
J66380.03
または、製品番号J66380-03
1 g
製品番号(カタログ番号) J66380.03
または、製品番号J66380-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7240-38-2
IUPAC Namesodium (2S,5R,6R)-3,3-dimethyl-6-(5-methyl-3-phenyl-1,2-oxazole-4-amido)-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrate
Molecular FormulaC19H20N3NaO6S
InChI KeyZVIYWUUZWWBNMB-VICXVTCVSA-M
SMILESO.[Na+].CC1=C(C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C(=NO1)C1=CC=CC=C1
さらに表示
Assay from Supplier's CofA815-950 µg/mg (dried basis)
Appearance (Color)White
FormPowder or crystals or crystalline powder
Water Content3.5-5.0%
Used primarily against Gram-positive organisms namely resistant Staphylococcus species

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used primarily against gram positive organisms namely resistant Staphylococcus species
RUO – Research Use Only