Neuropeptide Y (3-36), Human
Thermo Scientific Chemicals
Neuropeptide Y (3-36), Human
For cellular and molecular biology applications
製品番号(カタログ番号)数量
J66381.MCR
または、製品番号J66381-M^
1 mg
製品番号(カタログ番号) J66381.MCR
または、製品番号J66381-M^
価格(JPY)
-
数量:
1 mg
一括またはカスタム形式をリクエストする
仕様
フォーマットSolid
分子量4011.44
製品タイプNeuropeptide Y (3-36), human
精製方法HPLC
数量1 mg
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
組成および保存条件
-30°C to -10°C