CREBtide
Thermo Scientific Chemicals

CREBtide

For cellular and molecular biology applications
製品番号(カタログ番号) J66434.MCR
または、製品番号J66434-M^
価格(JPY)
-
数量:
1 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS149155-45-3
Appearance (Color)White
FormLyophilized Powder
Assay from Supplier's CofAPeptide content: Report Value
Assay from Supplier's CofAPeptide Purity: Report Value
Mass SpecExhibits correct MW
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
組成および保存条件
-30°C to -10°C

よくあるご質問(FAQ)

Is the CREBtide peptide acetylated at the N-terminus?

No, the CREBtide peptide is not acetylated at the N-terminus.

What is the amino acid sequence of the CREBtide peptide?

The amino acid sequence of the CREBtide peptide is: Lys-Arg-Arg-Glu-Ile-Leu-Ser-Arg-Arg-Pro-Ser-Tyr-Arg