Thermo Scientific Chemicals

Hydroxyapatite nanoparticles, 5-10% (w/v) aqueous colloidal dispersion, 20-50nm particles

Catalog number: J66691.AE
100 mL, Each
Thermo Scientific Chemicals

Hydroxyapatite nanoparticles, 5-10% (w/v) aqueous colloidal dispersion, 20-50nm particles

Catalog number: J66691.AE
100 mL, Each
Quantity
Catalog number: J66691.AE
also known as J66691-AE
Price (USD)
Price: 142.00
Online price: 119.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12167-74-7
IUPAC Name
tricalcium diphosphate
Molecular Formula
Ca3O8P2
InChI Key
QORWJWZARLRLPR-UHFFFAOYSA-H
SMILES
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Assay (unspecified)
Particle size: 20-50nm
Assay (unspecified)
Percent solids: 5-10%

Description

Hydroxyapatite nanoparticles is used for polishing and dental powders, porcelain and pottery, and enameling. It is also utilized to engineer new biomaterials for applications such as bone grafts and fillers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text