Colistin sodium methanesulfonate
Colistin sodium methanesulfonate
Colistin sodium methanesulfonate
Thermo Scientific Chemicals

Colistin sodium methanesulfonate

CAS: 8068-28-8 | C58H105N16Na5O28S5 | 1749.811 g/mol
製品番号(カタログ番号) J66698.06
または、製品番号J66698-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS8068-28-8
IUPAC Namepentasodium [(3-{3-hydroxy-2-[2-(6-methyloctanamido)-4-[(sulfonatomethyl)amino]butanamido]butanamido}-3-{[3-(1-hydroxyethyl)-12,15-bis(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-6,9,18-tris({2-[(sulfonatomethyl)amino]ethyl})-1,4,7,10,13,16,19-heptaazacyclotricosan-21-yl]carbamoyl}propyl)amino]methanesulfonate
Molecular FormulaC58H105N16Na5O28S5
InChI KeyIQWHCHZFYPIVRV-UHFFFAOYNA-I
SMILES[Na+].[Na+].[Na+].[Na+].[Na+].CCC(C)CCCCC(=O)NC(CCNCS([O-])(=O)=O)C(=O)NC(C(C)O)C(=O)NC(CCNCS([O-])(=O)=O)C(=O)NC1CCNC(=O)C(NC(=O)C(CCNCS([O-])(=O)=O)NC(=O)C(CCNCS([O-])(=O)=O)NC(=O)C(CC(C)C)NC(=O)C(CC(C)C)NC(=O)C(CCNCS([O-])(=O)=O)NC1=O)C(C)O
さらに表示
Heavy Metals≤ 0.003%
Loss on Drying≤ 5.0 %
pH6.5 to 8.5
Appearance (Color)White to off-white
FormPowder
さらに表示
Colistin sodium methanesulfate is used to permeabilize bacterial cell membranes and to study mannose-resistant haemagglutination. It is used to study effective treatments of P. aeruginosa infections in cystic fibrosis patients and to study the pharmacokinetics and BAL concentration of colistin in critically ill patients

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Freely soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

General References:

  1. Kimberly J. Griffitt.; D. Jay Grimes. A novel agar formulation for isolation and direct enumeration of Vibrio vulnificus from oyster tissue. Journal of Microbiological Methods. 2013, 94, (2), 98-102.
  2. Masamichi Fukuda.; Kazuyuki Sasaki. General Purpose Antimicrobial Ophthalmic Solutions Evaluated Using New Pharmacokinetic Parameter of Maximum Drug Concentration in Aqueous. Japanese Journal of Ophthalmology. 2002, 46, (4), 384-390.