Sulfamerazine sodium salt
Sulfamerazine sodium salt
Sulfamerazine sodium salt
Thermo Scientific Chemicals

Sulfamerazine sodium salt

CAS: 127-58-2 | C11H11N4NaO2S | 286.285 g/mol
製品番号(カタログ番号)数量パッケージ
J66717.30
または、製品番号J66717-30
250 gPlastic bottle
製品番号(カタログ番号) J66717.30
または、製品番号J66717-30
価格(JPY)
-
数量:
250 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS127-58-2
Appearance (Color)White to pale yellow
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only