Thermo Scientific Chemicals

Sodium glucoheptonate, Thermo Scientific Chemicals

Catalog number: J66811.22
100 g, Each
Thermo Scientific Chemicals

Sodium glucoheptonate, Thermo Scientific Chemicals

Catalog number: J66811.22
100 g, Each
Quantity
Catalog number: J66811.22
also known as J66811-22
Price (USD)
Price: 43.20
Online price: 36.65
Your price:
Quantity
-

Chemical Identifiers

CAS
31138-65-5
IUPAC Name
sodium (3R,4S,5R,6R)-2,3,4,5,6,7-hexahydroxyheptanoate
Molecular Formula
C7H13NaO8
InChI Key
FMYOMWCQJXWGEN-BMZZJELJSA-M
SMILES
[Na+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)C([O-])=O
Assay (unspecified)
98.5% min.
Chloride content
0.01% max.
Assay (unspecified)
Sulfate: 0.1% max.
Water Content (Karl Fischer Titration)
14.0% max.

Description

Sodium glucoheptonate is used for cleaning metal, mercerizing, paint stripping, and aluminum etching. It is also used to mercerize cotton, linen, hemp, and other plant-based fibers to increase strength and add luster to their overall appearance of threads and fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium glucoheptonate is used for cleaning metal, mercerizing, paint stripping, and aluminum etching. It is also used to mercerize cotton, linen, hemp, and other plant-based fibers to increase strength and add luster to their overall appearance of threads and fabrics.

Solubility
Freely soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text