Fibrinopeptide B, Human
Thermo Scientific Chemicals

Fibrinopeptide B, Human

For cellular and molecular biology applications
製品番号(カタログ番号) J66812.LB0
または、製品番号J66812-#0
価格(JPY)
-
数量:
5 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS36204-23-6
Appearance (Color)White
FormLyophilized powder
Assay (HPLC)≥98.0%
Assay (special)Peptide content >84%
Solubility1mg/ml in water
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
組成および保存条件
-30°C to -10°C