Thermo Scientific Chemicals

Sucrose stearate

Catalog number: J66845.14
25 g, Each
Thermo Scientific Chemicals

Sucrose stearate

Catalog number: J66845.14
25 g, Each
Quantity
Catalog number: J66845.14
also known as J66845-14
Price (USD)
Price: 121.00
Online price: 102.65
Your price:
Quantity
-

Chemical Identifiers

CAS
25168-73-4
IUPAC Name
2-{[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol; octadecanoic acid
Molecular Formula
C30H58O13
InChI Key
ONAIRGOTKJCYEY-UHFFFAOYNA-N
SMILES
CCCCCCCCCCCCCCCCCC(O)=O.OCC1OC(CO)(OC2OC(CO)C(O)C(O)C2O)C(O)C1O
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
Form
Powder
Assay (unspecified)
Monoester: Report value

Description

Sucrose stearate is used as an emulsifier in many beauty applications and cosmetics such as makeup products, cleansing formulas. Emulsifiers aid the blending of water and fatty substances, a common ingredient in many foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucrose stearate is used as an emulsifier in many beauty applications and cosmetics such as makeup products, cleansing formulas. Emulsifiers aid the blending of water and fatty substances, a common ingredient in many foods.

Solubility
Soluble in water and ethanol at 75°C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text