Cefoxitin, 98%
Cefoxitin, 98%
Cefoxitin, 98%
Thermo Scientific Chemicals

Cefoxitin, 98%

CAS: 35607-66-0 | C16H17N3O7S2 | 427.446 g/mol
製品番号(カタログ番号)数量
J66891.03
または、製品番号J66891-03
1 g
製品番号(カタログ番号) J66891.03
または、製品番号J66891-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS35607-66-0
IUPAC Name(6R,7S)-3-[(carbamoyloxy)methyl]-7-methoxy-8-oxo-7-[2-(thiophen-2-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Molecular FormulaC16H17N3O7S2
InChI KeyWZOZEZRFJCJXNZ-ZBFHGGJFSA-N
SMILESCO[C@]1(NC(=O)CC2=CC=CS2)[C@H]2SCC(COC(N)=O)=C(N2C1=O)C(O)=O
さらに表示
Appearance (Color)White to off-white
Mass SpecConforms
Purity≥97.5%
FormPowder
Cefoxitin is a semisynthetic, broad-spectrum second-generation cephalosporin with antibacterial activity. The activity of cefoxitin results in the weakening of the bacterial cell wall and causes cell lysis. Cefoxitin acts by interfering with cell wall synthesis. Its activity spectrum includes a broad range of gram-negative and gram-positive bacteria including anaerobes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cefoxitin is a semisynthetic, broad-spectrum second-generation cephalosporin with antibacterial activity. The activity of cefoxitin results in the weakening of the bacterial cell wall and causes cell lysis. Cefoxitin acts by interfe
RUO – Research Use Only
H. Knothe.; P. Shah.; V. Krcmery.; M. Antal.; S. Mitsuhashi. Transferable resistance to cefotaxime, cefoxitin, cefamandole and cefuroxime in clinical isolates of Klebsiella pneumoniae and Serratia marcescens.Infection. 1983, 11 (6),315-317 .