Fibronectin-Binding Protein
Thermo Scientific Chemicals

Fibronectin-Binding Protein

For research use
製品番号(カタログ番号) J66903.MCR
または、製品番号J66903-M^
価格(JPY)
-
数量:
1mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS119977-20-7
Assay from Supplier's CofAPeptide purity: ≥94.0% (HPLC)
Appearance (Color)White
FormLyophilized powder
Assay from Supplier's CofAPeptide Content: Report %
Mass SpecElectrospray exhibits correct molecular weight
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only