Thermo Scientific Chemicals
Gold nanoparticles, 100nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 572nm absorption, Thermo Scientific Chemicals
Catalog number: J67004.AC
25 mL, Each
Thermo Scientific Chemicals
Gold nanoparticles, 100nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 572nm absorption, Thermo Scientific Chemicals
Catalog number: J67004.AC
25 mL, Each
Quantity
Catalog number: J67004.AC
also known as J67004-AC
Price (USD)
Price: 111.00
Online price: 94.65
Your price:
Quantity
-
Description
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications