Thermo Scientific Chemicals

GDC-0068, 98%

Catalog number: J67082.LB0
5 mg, Each
Thermo Scientific Chemicals

GDC-0068, 98%

Catalog number: J67082.LB0
5 mg, Each
Quantity
Catalog number: J67082.LB0
also known as J67082-#0
Price (USD)

Chemical Identifiers

CAS
1001264-89-6
IUPAC Name
2-(4-chlorophenyl)-1-(4-{7-hydroxy-5-methyl-5H,6H,7H-cyclopenta[d]pyrimidin-4-yl}piperazin-1-yl)-3-[(propan-2-yl)amino]propan-1-one
Molecular Formula
C24H32ClN5O2
InChI Key
GRZXWCHAXNAUHY-UHFFFAOYNA-N
SMILES
CC(C)NCC(C(=O)N1CCN(CC1)C1=NC=NC2=C1C(C)CC2O)C1=CC=C(Cl)C=C1
Assay from Supplier's CofA
≥97.5% (HPLC)
Form
Powder
Mass Spec
Conforms
Appearance (Color)
White to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text