Gold nanoparticles, 150nm, supplied in 0.1mM PBS, 95%
Gold nanoparticles, 150nm, supplied in 0.1mM PBS, 95%
Gold nanoparticles, 150nm, supplied in 0.1mM PBS, 95%
Gold nanoparticles, 150nm, supplied in 0.1mM PBS, 95%
Thermo Scientific Chemicals

Gold nanoparticles, 150nm, supplied in 0.1mM PBS, 95%

CAS: 7440-57-5 | Au | 196.97 g/mol
製品番号(カタログ番号) J67276.AC
または、製品番号J67276-AC
価格(JPY)
-
お問い合わせください ›
数量:
25 mL
一括またはカスタム形式をリクエストする
仕様
CAS7440-57-5
ChEBICHEBI:30050
化学物質名または材質Gold nanoparticles
Brown to Pink
IUPAC名gold
さらに表示
For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
RUO – Research Use Only