TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade
TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade
TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade
Thermo Scientific Chemicals

TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade

Use directly to re-suspend and/or dilute purified DNA or RNA | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Nalgene bottle
Catalog number J75793.AP
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Quantity:
500 mL
Packaging:
Nalgene bottle
Request bulk or custom format
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Specifications
CAS1185-53-1
Chemical Name or MaterialTE Buffer
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
InChI KeyQKNYBSVHEMOAJP-UHFFFAOYSA-N
Molecular FormulaC4H12ClNO3
View more
Use directly to re-suspend and/or dilute purified DNA or RNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only