PureLink™ Pro 96 PCR Purification Kit - FAQs
PureLink™ Pro 96 PCR Purification Kit - FAQs
View additional product information for PureLink™ Pro 96 PCR Purification Kit - FAQs (K310096A)
4 product FAQs found
If I do not use all the wells in the filter plate of the PureLink Pro 96 Purification System, can I use those wells at another time?
Yes. Keep the unused wells covered with Foil Tape. Please note that extra Foil Tape and Receiver Plates will be needed.
My centrifuge does not reach 3,000 x g. For the PureLink Pro 96 system, could I spin at a lower force for a longer time?
Generally speaking, yes. It should work the same if you can keep the same g (rcf) x time.
What is the minimum and maximum size DNA fragments that can be isolated using the PureLink Pro 96 PCR Purification kit?
The kit facilitates routine purifications of double-stranded DNA fragments from 100 bp to 12 kb.
What downstream applications can the purified PCR product from PureLink Pro 96 PCR Purificaiton kit be used for?
The purified PCR product is suitable for automated fluorescent DNA sequencing, restriction enzyme digestion, cloning.