(2-Hydroxyethyl)triphenylphosphonium bromide, 98+%
(2-Hydroxyethyl)triphenylphosphonium bromide, 98+%
(2-Hydroxyethyl)triphenylphosphonium bromide, 98+%
Thermo Scientific Chemicals

(2-Hydroxyethyl)triphenylphosphonium bromide, 98+%

CAS: 7237-34-5 | C20H20BrOP | 387.26 g/mol
製品番号(カタログ番号) L00679.14
または、製品番号L00679-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7237-34-5
IUPAC Name(2-hydroxyethyl)triphenylphosphanium bromide
Molecular FormulaC20H20BrOP
InChI KeyQZJOQNHOOVSESC-UHFFFAOYSA-M
SMILES[Br-].OCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Assay (Titration ex Bromide)≥98.0 to ≤102.0%
Melting Point (clear melt)215-222?C
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Water Content (Karl Fischer Titration)<1.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only