Thermo Scientific Chemicals

Bromopyruvic acid, 97%, Thermo Scientific Chemicals

Catalog number: L00720.09
10 g, Each
Thermo Scientific Chemicals

Bromopyruvic acid, 97%, Thermo Scientific Chemicals

Catalog number: L00720.09
10 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L00720.09
also known as L00720-09
Price (USD)
Price: 99.10
Your price:
Quantity
-

Chemical Identifiers

CAS
1113-59-3
IUPAC Name
3-bromo-2-oxopropanoic acid
Molecular Formula
C3H3BrO3
InChI Key
PRRZDZJYSJLDBS-UHFFFAOYSA-N
SMILES
OC(=O)C(=O)CBr
Appearance (Color)
White to pale cream or pale orange
Form
Crystals or powder or crystalline powder and/or lumps
Assay (Titration ex Bromide)
≥96.0% to ≤104.0%
Comment
May contain some coloured lumpy solid from the manufacturing process
Infrared spectrum
Conforms

Description

Bromopyruvic acid is used as an affinity label for cysteine residues and a potential anti-cancer agent. It is involved in the synthesis of imidazo[1,2-a]pyridine-2-carboxylic acids and substituted pyranones. It plays a vital role as a cross-linker between nucleic acids and proteins. It is used as an anticancer agent for lung tumors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromopyruvic acid is used as an affinity label for cysteine residues and a potential anti-cancer agent. It is involved in the synthesis of imidazo[1,2-a]pyridine-2-carboxylic acids and substituted pyranones. It plays a vital role as a cross-linker between nucleic acids and proteins. It is used as an anticancer agent for lung tumors.

Solubility
Soluble in water, dimethyl sulfoxide, ethyl acetate and water.

Notes
Moisture and air sensitive. Store in cool place. Incompatible with strong oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text