Ethyl 5-hydroxyindole-2-carboxylate, 98+%
Ethyl 5-hydroxyindole-2-carboxylate, 98+%
Ethyl 5-hydroxyindole-2-carboxylate, 98+%
Ethyl 5-hydroxyindole-2-carboxylate, 98+%
Thermo Scientific Chemicals

Ethyl 5-hydroxyindole-2-carboxylate, 98+%

CAS: 24985-85-1 | C11H11NO3 | 205.213 g/mol
製品番号(カタログ番号) L00774.03
または、製品番号L00774-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS24985-85-1
FormCrystals or powder or crystalline powder or lumps or fused solid or granules
Assay (GC)≥98.0%
Appearance (Color)White to cream to brown or pink to gray
Assay (HPLC)≥98.0%
Ethyl 5-hydroxyindole-2-carboxylate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-hydroxyindole-2-carboxylate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Insoluble in water.

Notes
Store between 2-8 C. Store in cool, dryplace in a well ventilated area. Store in a tightly sealed container. Protect from moisture, heat.
RUO – Research Use Only