Appearance (Color)Pale cream to cream to pale brown or pink
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0% [UK sourced material]
Assay (Silylated GC)≥98.0% [UK sourced material]
FormCrystals or powder or crystalline powder or fused solid
Assay from Supplier's CofA≥98.0% (H-NMR) [US sourced material]
さらに表示