4-Ethyltoluene, 97%
4-Ethyltoluene, 97%
4-Ethyltoluene, 97%
Thermo Scientific Chemicals

4-Ethyltoluene, 97%

CAS: 622-96-8 | C9H12 | 120.195 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) L02120.14
または、製品番号L02120-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS622-96-8
IUPAC Name1-ethyl-4-methylbenzene
Molecular FormulaC9H12
InChI KeyJRLPEMVDPFPYPJ-UHFFFAOYSA-N
SMILESCCC1=CC=C(C)C=C1
さらに表示
Appearance (Color)Clear, colorless
FormLiquid
Refractive Index1.4925-1.4975 @ 20?C
Assay (GC)>96.0%
4-Ethyltoluene is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethyltoluene is used as intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
RUO – Research Use Only

General References:

  1. Consuelo Pizarro; Octavio Suárez-Iglesias; Ignacio Medina and Julio L. Bueno. Binary Diffusion Coefficients of 2-Ethyltoluene, 3-Ethyltoluene, and 4-Ethyltoluene in Supercritical Carbon Dioxide.J. Chem. Eng. Data.2009, 54 (5), 1467-1471.
  2. Tomas T; Swiercz R; Wiaderna D. Changes in function of rabbit cerebral cortex after intraperitoneal injections of organic solvents. II. Pseudocumene and 4-ethyltoluene.Medycyna Pracy.1997, 48 (3), 307-315.