4,4'-cis-Stilbenedicarboxylic acid, 95%
4,4'-cis-Stilbenedicarboxylic acid, 95%
4,4'-cis-Stilbenedicarboxylic acid, 95%
4,4'-cis-Stilbenedicarboxylic acid, 95%
Thermo Scientific Chemicals

4,4'-cis-Stilbenedicarboxylic acid, 95%

CAS: 133005-88-6 | C16H12O4 | 268.268 g/mol
製品番号(カタログ番号)数量
L04364.03
または、製品番号L04364-03
1 g
製品番号(カタログ番号) L04364.03
または、製品番号L04364-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS133005-88-6
IUPAC Name4-[2-(4-carboxyphenyl)ethenyl]benzoic acid
Molecular FormulaC16H12O4
InChI KeySBBQDUFLZGOASY-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=CC2=CC=C(C=C2)C(O)=O)C=C1
さらに表示
Assay (HPLC)≥94.0%
Appearance (Color)White to cream to yellow
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
FormPowder or granular powder or flakes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only