2-Chloro-1,3-dinitrobenzene, 98+%
2-Chloro-1,3-dinitrobenzene, 98+%
2-Chloro-1,3-dinitrobenzene, 98+%
Thermo Scientific Chemicals

2-Chloro-1,3-dinitrobenzene, 98+%

CAS: 606-21-3 | C6H3ClN2O4 | 202.55 g/mol
製品番号(カタログ番号) L04593.06
または、製品番号L04593-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS606-21-3
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Melting Point (clear melt)82.5-88.5°C
Appearance (Color)Yellow to orange to brown
2-Chloro-1,3-dinitrobenzene can be used as intermediates of dyes and pigments. Also used as solvents in HPLC, NMR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-1,3-dinitrobenzene can be used as intermediates of dyes and pigments. Also used as solvents in HPLC, NMR.

Solubility
Soluble in alcohol. Moderately soluble in benzene, ether. Practically insoluble in water.

Notes
Incompatible materials are oxidizing agents, Strong bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
Ryun Youn Sun;, Hojune Choi; Jong Pal Lee; Jong Keun Park; Kiyull Yang; and In Sun Koo. Kinetic Studies on the Nucleophilic Substitution Reaction of 4-X-Substituted-2,6-dinitrochlorobenzene with Pyridines in MeOH-MeCN Mixtures.Bull. Korean Chem. Soc.2009, 30 (7), 1579-1582.