Thermo Scientific Chemicals

4-Bromoheptane, 97%, Thermo Scientific Chemicals

Catalog number: L04807.06
5 g, Each
Thermo Scientific Chemicals

4-Bromoheptane, 97%, Thermo Scientific Chemicals

Catalog number: L04807.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L04807.06
also known as L04807-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
998-93-6
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4440-1.4505 @ 20?C

Description

4-Bromoheptane is used in the preparation of 4-nitro-heptane by reacting with sodium nitrite as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoheptane is used in the preparation of 4-nitro-heptane by reacting with sodium nitrite as a reagent.

Solubility
Miscible with carbon tetrachloride, chloroform and benzene. Immiscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text