2,4,6-Trimethoxybenzoic acid, 98%
2,4,6-Trimethoxybenzoic acid, 98%
2,4,6-Trimethoxybenzoic acid, 98%
Thermo Scientific Chemicals

2,4,6-Trimethoxybenzoic acid, 98%

CAS: 570-02-5 | C10H12O5 | 212.20 g/mol
製品番号(カタログ番号) L04997.14
または、製品番号L04997-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS570-02-5
IUPAC Name2,4,6-trimethoxybenzoic acid
Molecular FormulaC10H12O5
InChI KeyJATAKEDDMQNPOQ-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=C(C(O)=O)C(OC)=C1
さらに表示
Assay (Silylated GC)≥97.5%
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only