Ethyl benzoylacetate, 90+%
Ethyl benzoylacetate, 90+%
Ethyl benzoylacetate, 90+%
Thermo Scientific Chemicals

Ethyl benzoylacetate, 90+%

CAS: 94-02-0 | C11H12O3 | 192.214 g/mol
数量:
50 g
250 g
1000 g
製品番号(カタログ番号) L05030.18
または、製品番号L05030-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS94-02-0
IUPAC Nameethyl 3-oxo-3-phenylpropanoate
Molecular FormulaC11H12O3
InChI KeyGKKZMYDNDDMXSE-UHFFFAOYSA-N
SMILESCCOC(=O)CC(=O)C1=CC=CC=C1
さらに表示
Assay (HPLC)≥90.0%
Appearance (Color)Clear colorless to white to yellow to orange to brown
Refractive Index1.4875-1.5675 @ 20?C
FormLiquid
Ethyl benzoylacetate is used as a flavoring agent and an intermediate in organic synthesis. Further, it is used in the preparation of triazipinones, iodonium ylides, ethyl 2-fluoro-2-benzolyacetate and benzo[c]furan-4,7-diones. It is involved in the Clisen condensation reaction with malononitrile to prepare 2-cyano-5-phenyl-3,5-dioxopentanonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl benzoylacetate is used as a flavoring agent and an intermediate in organic synthesis. Further, it is used in the preparation of triazipinones, iodonium ylides, ethyl 2-fluoro-2-benzolyacetate and benzo[c]furan-4,7-diones. It is involved in the Clisen condensation reaction with malononitrile to prepare 2-cyano-5-phenyl-3,5-dioxopentanonitrile.

Solubility
Miscible with ethanol and ether. Immiscible with water.

Notes
Air and light sensitive. Incompatible with acids, bases, oxidizing agents and reducing agents.
RUO – Research Use Only

General References:

  1. Has been used in a one-pot synthesis of ɑ-substituted acrylic esters: Synth. Commun., 18, 1213 (1988):
  2. Compare also Ethyl acetoacetate, A12544.
  3. Rajesh, N.; Prajapati, D. Indium-catalyzed, novel route to beta,beta-disubstituted indanones via tandem Nakamura addition-hydroarylation-decarboxylation sequence. Chem. Commun. 2015, 51 (16), 3347-3350.
  4. Ni, Q.; Xiong, J.; Song, X.; Raabe, G.; Enders, D. NHC-catalyzed activation of alpha,beta-unsaturated N-acyltriazoles: an easy access to dihydropyranones. Chem. Commun. 2015, 51 (78), 14628-14631.