Thermo Scientific Chemicals

Citraconic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: L05238.14
25 g, Each
Thermo Scientific Chemicals

Citraconic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: L05238.14
25 g, Each
Quantity
Catalog number: L05238.14
also known as L05238-14
Price (USD)
Price: 36.10
Online price: 30.65
Your price:
Quantity
-

Chemical Identifiers

CAS
616-02-4
IUPAC Name
3-methyl-2,5-dihydrofuran-2,5-dione
Molecular Formula
C5H4O3
InChI Key
AYKYXWQEBUNJCN-UHFFFAOYSA-N
SMILES
CC1=CC(=O)OC1=O
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4695-1.4740 @ 20?C

Description

Citraconic anhydride is used as a monomer for specialty unsaturated polyester resins. It is used to prepare bicyclic pyrrolidines, maleimides and co- and terpolymers. It is also used for the protection of N-terminal amino acids. Further, it is involved in the preparation of clothing ping acid and its amide. In addition to this, it is selectively used for the isolation of histidyl peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citraconic anhydride is used as a monomer for specialty unsaturated polyester resins. It is used to prepare bicyclic pyrrolidines, maleimides and co- and terpolymers. It is also used for the protection of N-terminal amino acids. Further, it is involved in the preparation of clothing ping acid and its amide. In addition to this, it is selectively used for the isolation of histidyl peptides.

Solubility
Miscible with water, alcohol, acetone and ether.

Notes
Moisture sensitive. Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text