alpha-Cyclohexylphenylacetonitrile, 98%
alpha-Cyclohexylphenylacetonitrile, 98%
alpha-Cyclohexylphenylacetonitrile, 98%
Thermo Scientific Chemicals

alpha-Cyclohexylphenylacetonitrile, 98%

CAS: 3893-23-0 | C14H17N | 199.30 g/mol
製品番号(カタログ番号) L05350.18
または、製品番号L05350-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3893-23-0
Appearance (Color)White to cream to yellow
Melting Point (clear melt)49-58?C
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or fused solid or lumps
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only