Thermo Scientific Chemicals

Molecular sieves, 4A, 3-5mm (0.12-0.20in) beads, Thermo Scientific Chemicals

Catalog number: L05466.0I
5000 g, Each
Thermo Scientific Chemicals

Molecular sieves, 4A, 3-5mm (0.12-0.20in) beads, Thermo Scientific Chemicals

Catalog number: L05466.0I
5000 g, Each
Quantity
Catalog number: L05466.0I
also known as L05466-0I
Price (USD)
Price: 302.00
Online price: 256.65
Your price:
Quantity
-

Chemical Identifiers

CAS
70955-01-0
IUPAC Name
Molecular sieves
Synonym
2-[2-acetamido-3-(3-fluorophenyl)propanamido]-5-methoxy-4-(methoxycarbonyl)-3-methyl-5-oxopentanoic acid
MDL Number
MFCD00131613
Comment
Water absorbtion >20% @ 50%rH, 20?C, 24h
Residual water
≤1.5% w/w @ 550?C
Particle Size
≤5% is < 2.5mm
Form
Beads
Particle Size
≤5% is > 5mm

Description

Molecular sieves, 4A, 3-5mm (0.12-0.20in) beads are useful in general gas drying applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text