Thermo Scientific Chemicals

N-Ethylbenzylamine, 95%, Thermo Scientific Chemicals

Catalog number: L05651.AE
100 mL, Each
Thermo Scientific Chemicals

N-Ethylbenzylamine, 95%, Thermo Scientific Chemicals

Catalog number: L05651.AE
100 mL, Each
Quantity
Catalog number: L05651.AE
also known as L05651-AE
Price (USD)
Price: 84.90
Online price: 72.65
Your price:
Quantity
-

Chemical Identifiers

CAS
14321-27-8
IUPAC Name
benzyl(ethyl)amine
Molecular Formula
C9H13N
InChI Key
HVAAHUDGWQAAOJ-UHFFFAOYSA-N
SMILES
CCNCC1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Assay (Non-aqueous acid-base Titration)
≥94.0 to ≤106.0%
Form
Liquid
Refractive Index
1.5090-1.5140 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text