5-Nonanol, 98%
5-Nonanol, 98%
5-Nonanol, 98%
Thermo Scientific Chemicals

5-Nonanol, 98%

CAS: 623-93-8 | C9H20O | 144.258 g/mol
製品番号(カタログ番号)数量
L05812.14
または、製品番号L05812-14
25 g
製品番号(カタログ番号) L05812.14
または、製品番号L05812-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
仕様
バイルシュタイン1733552
CAS623-93-8
化学物質名または材質5-Nonanol
密度0.826
EINECS番号210-820-2
さらに表示
5-Nonanol is used as intermediates in chemical research. It is used as natural substances and extractives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nonanol is used as intermediates in chemical research. It is used as natural substances and extractives.

Solubility
Soluble in water (0.46 mg/ml at 15°C), and chloroform.

Notes
Incompatible ma
RUO – Research Use Only