Methyl undecanoate, 99%
Methyl undecanoate, 99%
Methyl undecanoate, 99%
Thermo Scientific Chemicals

Methyl undecanoate, 99%

CAS: 1731-86-8 | C12H24O2 | 200.322 g/mol
製品番号(カタログ番号) L05939.06
または、製品番号L05939-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1731-86-8
IUPAC Namemethyl undecanoate
Molecular FormulaC12H24O2
InChI KeyXPQPWPZFBULGKT-UHFFFAOYSA-N
SMILESCCCCCCCCCCC(=O)OC
さらに表示
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4280-1.4320 @ 20?C
Assay (GC)≥98.5%
Methyl undecanoate is used as plasticizer in polymers. It is used as fragrance agent in aroma and flavor Chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl undecanoate is used as plasticizer in polymers. It is used as fragrance agent in aroma and flavor Chemicals.

Solubility
Soluble in ethanol, and ether. Insoluble in water.

Notes
Store at -20°C. Incompatible with oxidizing agents.
RUO – Research Use Only
Xinhai Yu; Zhenzhong Wen; Hongliang Li; Shan-Tung Tu; Jinyue Yan. Transesterification of Pistacia chinensis oil for biodiesel catalyzed by CaO-CeO2 mixed oxides. Fuel.2011, 90, 1868-1874.