n-Octyl gallate, 98+%
n-Octyl gallate, 98+%
n-Octyl gallate, 98+%
Thermo Scientific Chemicals

n-Octyl gallate, 98+%

CAS: 1034-01-1 | C15H22O5 | 282.336 g/mol
製品番号(カタログ番号) L06081.14
または、製品番号L06081-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1034-01-1
IUPAC Nameoctyl 3,4,5-trihydroxybenzoate
Molecular FormulaC15H22O5
InChI KeyNRPKURNSADTHLJ-UHFFFAOYSA-N
SMILESCCCCCCCCOC(=O)C1=CC(O)=C(O)C(O)=C1
さらに表示
Appearance (Color)White to pale cream
Assay (HPLC)≥98.0%
Water Content (Karl Fischer Titration)≤2.0%
Identification (FTIR)Conforms
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only