Methyl 2-nitro-3,4,5-trimethoxybenzoate, 98%
Methyl 2-nitro-3,4,5-trimethoxybenzoate, 98%
Methyl 2-nitro-3,4,5-trimethoxybenzoate, 98%
Thermo Scientific Chemicals

Methyl 2-nitro-3,4,5-trimethoxybenzoate, 98%

CAS: 5081-42-5 | C11H13NO7 | 271.225 g/mol
製品番号(カタログ番号) L06188.09
または、製品番号L06188-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5081-42-5
IUPAC Namemethyl 3,4,5-trimethoxy-2-nitrobenzoate
Molecular FormulaC11H13NO7
InChI KeyHCEIEWYPDCDNNU-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(OC)=C(OC)C(OC)=C1[N+]([O-])=O
さらに表示
Appearance (Color)White to cream or pale yellow
FormCrystals or crystalline powder or powder
Assay (GC)≥97.5%
Melting Point (clear melt)66-70°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only