cis-4-Decenal, 95%
cis-4-Decenal, 95%
cis-4-Decenal, 95%
Thermo Scientific Chemicals

cis-4-Decenal, 95%

CAS: 21662-09-9 | C10H18O | 154.253 g/mol
製品番号(カタログ番号) L06382.03
または、製品番号L06382-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS21662-09-9
IUPAC Name(4E,6E)-deca-4,6-dien-1-ol
Molecular FormulaC10H18O
InChI KeyFJHNCSRGAAVOQK-YTXTXJHMSA-N
SMILESCCCC=CC=CCCCO
さらに表示
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥94.0%
Refractive Index1.4410-1.4470 @ 20?C
FormLiquid
Free acid (titration)≤2.0%
cis-4-Decenal is suitable for use as standard compound to investigate the release of odorants in hydrocolloid model systems containing original or regio-selectively carboxylated cellulose at different pH values using static headspace gas chromatographic (SHS-GC) analysis. It may be used as standard for the standardization of 9-decenal by gas-chromatography. It may be used in the preparation of cis-4-decenol. It has been identified as one of the volatile biological marker for grey mold (Botrytis cinerea) infections in strawberry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-Decenal is suitable for use as standard compound to investigate the release of odorants in hydrocolloid model systems containing original or regio-selectively carboxylated cellulose at different pH values using static headspace gas chromatographic (SHS-GC) analysis. It may be used as standard for the standardization of 9-decenal by gas-chromatography. It may be used in the preparation of cis-4-decenol. It has been identified as one of the volatile biological marker for grey mold (Botrytis cinerea) infections in strawberry.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Thomas Vandendriessche; Johan Keulemans; Annemie Geeraerd; Bart M.Nicolai; Maarten L.A.T.M. Hertog. Evaluation of fast volatile analysis for detection of Botrytis cinerea infections in strawberry. Food Microbiology. 2012, 32,(2), 406-414.
  2. X.P.Luo; M.Yazdanpanah; N.Bhooi, D.C.Lehotay. Determination of aldehydes and other lipid peroxidation products in biological samples by gas chromatography-mass spectrometry. Analytical Biochemistry. 1995, 228,(2), 294-298.