4-tert-Butylbenzophenone, 98%
4-tert-Butylbenzophenone, 98%
4-tert-Butylbenzophenone, 98%
4-tert-Butylbenzophenone, 98%
Thermo Scientific Chemicals

4-tert-Butylbenzophenone, 98%

CAS: 22679-54-5 | C17H18O | 238.33 g/mol
製品番号(カタログ番号) L06386.03
または、製品番号L06386-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
バイルシュタイン2100075
CAS22679-54-5
化学物質名または材質4-tert-Butylbenzophenone
EINECS番号245-154-1
引火点150°C (302°F)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only