Glycolaldehyde dimethyl acetal, 98%, stab. with ca 0.1% sodium carbonate
Glycolaldehyde dimethyl acetal, 98%, stab. with ca 0.1% sodium carbonate
Glycolaldehyde dimethyl acetal, 98%, stab. with ca 0.1% sodium carbonate
Glycolaldehyde dimethyl acetal, 98%, stab. with ca 0.1% sodium carbonate
Thermo Scientific Chemicals

Glycolaldehyde dimethyl acetal, 98%, stab. with ca 0.1% sodium carbonate

CAS: 30934-97-5 | C4H10O3 | 106.12 g/mol
製品番号(カタログ番号) L07099.03
または、製品番号L07099-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS30934-97-5
IUPAC Name2,2-dimethoxyethan-1-ol
Molecular FormulaC4H10O3
InChI KeyNYPNCQTUZYWFGG-UHFFFAOYSA-N
SMILESCOC(CO)OC
さらに表示
Appearance (Color)Clear or slightly turbid, colorless to pale yellow
CommentProduct may appear slightly turbid or contain sediment upon storage due to presence of stabiliser.
Assay (GC)>97.5%
FormLiquid
Refractive Index1.4110-1.4150 @ 20?C
Glycolaldehyde dimethyl acetal is used in the preparation of aceto- xyacetaldehyde dimethyl acetal by reacting with acetic anhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycolaldehyde dimethyl acetal is used in the preparation of aceto- xyacetaldehyde dimethyl acetal by reacting with acetic anhydride.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Melappa, G.; Naik, C. Determination of Phytochemicals by GC-MS in two Fractions (17 and 21) of Methanol Extract of Loranthus micranthus and their Antioxidant and Anti-inflammatory Activity. J. Nat. Prod. 2015, 5 (3), 158-175.
  2. Ventosa-Andrés, P.; La-Venia, A.; Ripoll, C. A. B.; Hradilová, L.; Krchňák, V. Synthesis of Nature-Inspired Medium-Sized Fused Heterocycles from Amino Acids. Chem. Eur. J. 2015, 21 (37), 13112-13119.