Methyl 4-chloro-3-nitrobenzoate, 98%
Methyl 4-chloro-3-nitrobenzoate, 98%
Methyl 4-chloro-3-nitrobenzoate, 98%
Methyl 4-chloro-3-nitrobenzoate, 98%
Thermo Scientific Chemicals

Methyl 4-chloro-3-nitrobenzoate, 98%

CAS: 14719-83-6 | C8H6ClNO4 | 215.59 g/mol
製品番号(カタログ番号) L07407.14
または、製品番号L07407-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14719-83-6
IUPAC Namemethyl 4-chloro-3-nitrobenzoate
Molecular FormulaC8H6ClNO4
InChI KeyXRTKWPWDSUNLHS-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
さらに表示
Appearance (Color)White to pale yellow to cream
Melting Point (clear melt)77.5-83.5?C
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Methyl 4-chloro-3-nitrobenzoate, is used as starting material for the preparation of the methyl benzoates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-chloro-3-nitrobenzoate, is used as starting material for the preparation of the methyl benzoates

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. M. K. ChantooniJr.; I. M. Kolthoff. Resolution of acid strength in tert-butyl alcohol and isopropyl alcohol of substituted benzoic acids, phenols, and aliphatic carboxylic acids. Anal. Chem. 1979, 51, (1), 133-140.
  2. Joseph Miller. The SN Mechanism in Aromatic Compounds. VI. Carbonyl and Nitrile Substituents. J. Am. Chem. Soc. 1954, 76, (2), 448-450.