Thermo Scientific Chemicals

4-Dimethylamino-1-naphthoic acid, 98%, Thermo Scientific Chemicals

5 g, Each
Thermo Scientific Chemicals

4-Dimethylamino-1-naphthoic acid, 98%, Thermo Scientific Chemicals

5 g, Each
Quantity
Catalog number: L07944.06
also known as L07944-06
Price (USD)
Price: 455.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
78062-03-0
IUPAC Name
4-(dimethylamino)naphthalene-1-carboxylic acid
Molecular Formula
C13H13NO2
InChI Key
LZQKNRRVTOJUHQ-UHFFFAOYSA-N
SMILES
CN(C)C1=C2C=CC=CC2=C(C=C1)C(O)=O
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
White to cream to yellow to pale brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only