1,2,3,4-Tetrahydroisoquinoline, 97%
1,2,3,4-Tetrahydroisoquinoline, 97%
1,2,3,4-Tetrahydroisoquinoline, 97%
Thermo Scientific Chemicals

1,2,3,4-Tetrahydroisoquinoline, 97%

CAS: 91-21-4 | C9H11N | 133.19 g/mol
製品番号(カタログ番号) L08143.14
または、製品番号L08143-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS91-21-4
Refractive Index1.5655-1.5710 @ 20°C
Appearance (Color)Clear colorless to yellow
FormLiquid or viscous liquid
Assay (GC)≥95.0%
It is used as a reagent in the preparation of4-(1,2,4-oxadiazol-5-yl)piperidine-1-carboxamides as antiproliferative tubulin inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent in the preparation of4-(1,2,4-oxadiazol-5-yl)piperidine-1-carboxamides as antiproliferative tubulin inhibitors.

Solubility
Soluble in water at 20°C 20g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
RUO – Research Use Only

General References:

  1. Hiroshi Kawai, et al. Novel Endogenous 1,2,3 ,4-Tetrahydroisoquinoline Derivatives: Uptake by Dopamine Transporter and Activity to Induce Parkinsonism.J. Neurochem.,1998,70(2), 745-751.
  2. Estefanía Méndez-Álvarez, et al. Inhibition of brain monoamine oxidase by adducts of 1,2,3,4-tetrahydroisoquinoline with components of cigarette smoke.Life Sci.,1997,60(19), 1719-1727.