Methyl 4-cyanobenzoate, 99%
Methyl 4-cyanobenzoate, 99%
Methyl 4-cyanobenzoate, 99%
Methyl 4-cyanobenzoate, 99%
Thermo Scientific Chemicals

Methyl 4-cyanobenzoate, 99%

CAS: 1129-35-7 | C9H7NO2 | 161.16 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) L08365.22
または、製品番号L08365-22
価格(JPY)
-
見積もりを依頼する
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1129-35-7
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White to cream to pale yellow
Assay (GC)≥98.5%
It finds its application in the chemoselective biotransformation of nitriles by Rhodococcus equi A4 and in the design of 3-(4-carboxybenzoyl)-2-quinolinecarboxaldehyde as a reagent for ultrasensitive determination of primary amines by capillary electrophoresis using laser fluorescence detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the chemoselective biotransformation of nitriles by Rhodococcus equi A4 and in the design of 3-(4-carboxybenzoyl)-2-quinolinecarboxaldehyde as a reagent for ultrasensitive determination of primary amines by capillary electrophoresis using laser fluorescence detection.

Solubility
Insoluble in water. Soluble in chloroform and ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Kevin McMahon, et al. The photochemical nucleophile-olefin combination, aromatic substitution (photo-NOCAS) reaction (Part 4): methanol-olefins, methyl 4-cyanobenzoate.Can. J. Chem.,1993,71(4), 450-468.
  2. Ludmila Martínková, et al. Chemoselective biotransformation of nitriles by Rhodococcus equi A4.Biotechnol. Lett.,1998,20(10), 909-912.